Product Details

SNP ID
rs33300284
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.5:130212393 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGCCTGGTGCCCATGCCTATAGTCT[C/T]AGGACTCTAGAAGCAGAGGTACCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Rabgef1 PubMed Links

Gene Details

Gene
Rabgef1
Gene Name
RAB guanine nucleotide exchange factor (GEF) 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199059.1 Intron NP_001185988.1
NM_019983.2 Intron NP_064367.1
Gene
Tmem248
Gene Name
transmembrane protein 248
There are no transcripts associated with this gene.

View Full Product Details