Product Details

SNP ID
rs13480892
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:6624227 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCATCAGGCGAACGGCCATGATGCT[A/G]GGCGCAGGGAAGAGGGAGGGAGGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Snora5c PubMed Links

Gene Details

Gene
Snora5c
Gene Name
small nucleolar RNA, H/ACA box 5C
There are no transcripts associated with this gene.

Gene
Tbrg4
Gene Name
transforming growth factor beta regulated gene 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130457.1 225 UTR 5 NP_001123929.1
NM_134011.2 225 UTR 5 NP_598772.1

View Full Product Details