Product Details
- SNP ID
-
rs6268584
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:151850979 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGGGCTCATCCCAGACCTACAAG[A/G]TGTGAACTGCATACCTCATTTGCGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Rspo4
PubMed Links
Gene Details
- Gene
- Rspo4
- Gene Name
- R-spondin family, member 4
View Full Product Details