Product Details

SNP ID
rs13482698
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:89165784 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGGACTTCTCCCGGCTCCACAGCCA[A/G]TGTCCGGTTTCCTCGGCCCGTGAGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Mir7118 PubMed Links

Gene Details

Gene
Mir7118
Gene Name
microRNA 7118
There are no transcripts associated with this gene.

Gene
Plxnb2
Gene Name
plexin B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159521.2 1769 Silent Mutation NP_001152993.1
NM_001284506.1 1769 Silent Mutation NP_001271435.1
NM_138749.3 1769 Silent Mutation NP_620088.2

View Full Product Details