Product Details

SNP ID
rs13475977
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:90612202 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTGCCCCAGTTCCGCCAATCCCCAA[C/T]GAGCAGCAGCTCGCCCGGCTCACCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Cops8 PubMed Links

Gene Details

Gene
Cops8
Gene Name
COP9 (constitutive photomorphogenic) homolog, subunit 8 (Arabidopsis thaliana)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133805.3 704 Silent Mutation NP_598566.3

View Full Product Details