Product Details
- SNP ID
-
rs13482134
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:34735571 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAAAAAATAGTGGATTTGATGGT[C/T]GAAAGATGCCTTGGCAGCTGGAGCG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Wapal
PubMed Links
Gene Details
- Gene
- Wapal
- Gene Name
- wings apart-like homolog (Drosophila)
There are no transcripts associated with this gene.
View Full Product Details