Product Details

SNP ID
rs13483567
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.19:22978380 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CAGGAGCATGGATTGTCCCGGCCAT[A/C]ATGGCCTGCTACCTCTTGGTGGCGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Trpm3 PubMed Links

Gene Details

Gene
Trpm3
Gene Name
transient receptor potential cation channel, subfamily M, member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035239.2 3254 Silent Mutation NP_001030316.1
NM_001035240.2 3254 Silent Mutation NP_001030317.1
NM_001035241.2 3254 Silent Mutation NP_001030318.1
NM_001035242.1 3254 Silent Mutation NP_001030319.1
NM_001035243.2 3254 Silent Mutation NP_001030320.1
NM_001035244.1 3254 Intron NP_001030321.1
NM_001035245.1 3254 Intron NP_001030322.1
NM_001035246.1 3254 Intron NP_001030323.1
NM_177341.4 3254 Silent Mutation NP_796315.2

View Full Product Details