Product Details

SNP ID
rs13478092
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.5:3595407 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGGCTCCCACTGCAGGAACTCCTTG[A/T]ATTAGTAAGTACTGAGACTCCAAAT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Pex1 PubMed Links

Gene Details

Gene
Pex1
Gene Name
peroxisomal biogenesis factor 1
There are no transcripts associated with this gene.

Gene
Rbm48
Gene Name
RNA binding motif protein 48
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172991.4 621 Silent Mutation NP_766579.3

View Full Product Details