Product Details

SNP ID
rs13476309
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:192225810 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTGGTGGTTATATAGGTGTTGACTC[A/G]AGAGCTGCACTCTGGCCAGGAGCAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Kcnh1 PubMed Links

Gene Details

Gene
Kcnh1
Gene Name
potassium voltage-gated channel, subfamily H (eag-related), member 1
There are no transcripts associated with this gene.

View Full Product Details