Product Details

SNP ID
rs13481220
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:108519915 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCCTAGACATCTGGTGTGATTGAAG[A/G]GTTAGAACAGAACATCTGTCAATTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Cep112 PubMed Links

Gene Details

Gene
Cep112
Gene Name
centrosomal protein 112
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_029586.2 1721 Intron NP_083862.1
NM_029606.3 1721 Missense Mutation NP_083882.3
NM_145688.2 1721 Intron NP_663721.2

View Full Product Details