Product Details

SNP ID
rs6268898
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:81154152 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ACCCAAATTTGCCAATTGTGGTTTC[C/T]TTTCCCCCAGCCGTGTTTTGTATAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Nyap2 PubMed Links

Gene Details

Gene
Nyap2
Gene Name
neuronal tyrosine-phophorylated phosphoinositide 3-kinase adaptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172849.3 Intron NP_766437.2

View Full Product Details