Product Details
- SNP ID
-
rs13480654
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:75148046 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGTCTGTGCTCTCTAAGGGCAGC[A/G]ACCCTCCACTGGGCCATGGTGAAGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Bcr
PubMed Links
Gene Details
- Gene
- Bcr
- Gene Name
- breakpoint cluster region
View Full Product Details