Product Details

SNP ID
rs48479876
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:72834418 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AAGCCTAGTGCTACCAAGGGAACTC[G/T]GGGTGGCATGTTTGGGATGCTGAAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Zzef1 PubMed Links

Gene Details

Gene
Zzef1
Gene Name
zinc finger, ZZ-type with EF hand domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001045536.2 Intron NP_001039001.1

View Full Product Details