Product Details

SNP ID
rs13482505
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:30637218 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GAAGTGAAGGACAGAAGATAAACAG[A/C]CGGGCCCTTTTTCAGGATGCAGTGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Ctnnd2 PubMed Links

Gene Details

Gene
Ctnnd2
Gene Name
catenin (cadherin associated protein), delta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_008729.2 Intron NP_032755.2

View Full Product Details