Product Details

SNP ID
rs13481551
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:79079873 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TAGAGACAGGGATGGCCCACAGTTA[C/T]CAGGGCAATGGTACTGTATTTGGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
9230116L04Rik PubMed Links

Gene Details

Gene
9230116L04Rik
Gene Name
RIKEN cDNA 9230116L04 gene
There are no transcripts associated with this gene.

Gene
Pigh
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_029988.2 4657 Intron NP_084264.1
Gene
Plekhh1
Gene Name
pleckstrin homology domain containing, family H (with MyTH4 domain) member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181073.3 4657 UTR 3 NP_851418.2

View Full Product Details