Product Details
- SNP ID
-
rs6256981
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:47991663 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTGTGGGGGAAAGAATTAAGGTTT[A/G]TATAAAGGAGGTCGTGCTCCCATAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Csmd3
PubMed Links
Gene Details
- Gene
- Csmd3
- Gene Name
- CUB and Sushi multiple domains 3
View Full Product Details