Product Details

SNP ID
rs3693123
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.14:14741777 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TTTCTCTTTTAAGCCACACTTACTA[G/A]TAGTACCAGATATTTTAAGTATAAA
Phenotype
Polymorphism
G/A, Transition Substitution
Allele Nomenclature
Literature Links
Slc4a7 PubMed Links

Gene Details

Gene
Slc4a7
Gene Name
solute carrier family 4, sodium bicarbonate cotransporter, member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033270.2 Intron NP_001028442.2

View Full Product Details