Product Details

SNP ID
rs13482067
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.14:14765603 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GTGCAATAGAGTCTCTTTTTGGAGC[A/G]TCATTAACTGGGATTGCCTATTCAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Slc4a7 PubMed Links

Gene Details

Gene
Slc4a7
Gene Name
solute carrier family 4, sodium bicarbonate cotransporter, member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033270.2 1792 Silent Mutation NP_001028442.2

View Full Product Details