Product Details

SNP ID
rs4182165
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.16:45980323 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GCTTGTTCCCAAAGGCCTCCTGGAT[C/T]CGCAGAACCAGGCATTTGTGGTGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Plcxd2 PubMed Links

Gene Details

Gene
Plcxd2
Gene Name
phosphatidylinositol-specific phospholipase C, X domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134480.1 1094 Silent Mutation NP_001127952.1

View Full Product Details