Product Details
- SNP ID
-
rs13476269
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:180866166 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGAGGCTCAAGTGTGAACTGATGG[C/T]CCTTGGACTGAAGTGTGGGGGGACC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Sde2
PubMed Links
Gene Details
- Gene
- Sde2
- Gene Name
- SDE2 telomere maintenance homolog (S. pombe)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_145943.1 |
1242 |
Missense Mutation |
|
|
NP_666055.1 |
View Full Product Details