Product Details

SNP ID
rs13459122
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:80795365 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GTCTTGTGATGACCTCTGTACTTAA[A/T]GGGTGCATTGTTCTCTTCCCTTGGT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Amh PubMed Links

Gene Details

Gene
Amh
Gene Name
anti-Mullerian hormone
There are no transcripts associated with this gene.

Gene
Dot1l
Gene Name
DOT1-like, histone H3 methyltransferase (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199322.1 Intron NP_955354.1
Gene
Plekhj1
Gene Name
pleckstrin homology domain containing, family J member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023900.2 Intron NP_076389.2
Gene
Sf3a2
Gene Name
splicing factor 3a, subunit 2
There are no transcripts associated with this gene.

View Full Product Details