Product Details
- SNP ID
-
rs13475814
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:36879483 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGACTGCTAGAGTATTGATAACTT[C/T]TTATACACAATGGAGAATGAGTGTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Tmem131
PubMed Links
Gene Details
- Gene
- Tmem131
- Gene Name
- transmembrane protein 131
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018872.2 |
|
Intron |
|
|
NP_061360.2 |
View Full Product Details