Product Details
- SNP ID
-
rs4161847
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:11433756 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGAAAGGCATTAGGGCGCCAGGA[C/T]CCCTTCAAGGCATGTGCCTTGGTTA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Snx29
PubMed Links
Gene Details
- Gene
- Snx29
- Gene Name
- sorting nexin 29
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_028964.4 |
|
Intron |
|
|
NP_083240.2 |
View Full Product Details