Product Details

SNP ID
rs13459104
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.8:84662600 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GAACTCGGGCTGGTGGGCTTCTACC[G/T]TGGCTGAGAGGTAGACCTCTCGAGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Gm26664 PubMed Links

Gene Details

Gene
Gm26664
Gene Name
predicted gene, 26664
There are no transcripts associated with this gene.

Gene
Ier2
Gene Name
immediate early response 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_010499.4 253 Missense Mutation NP_034629.3
Gene
Nacc1
Gene Name
nucleus accumbens associated 1, BEN and BTB (POZ) domain containing
There are no transcripts associated with this gene.

View Full Product Details