Product Details
- SNP ID
-
rs13481968
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:98858290 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGTAAGTGATTTTCAAAGGTGGAAA[A/C]GCTAACGGTTCATACTTCACTTTGT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Tnpo1
PubMed Links
Gene Details
- Gene
- Tnpo1
- Gene Name
- transportin 1
View Full Product Details