Product Details
- SNP ID
-
rs6263895
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:30056734 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAACCCTGCTTCCATACATACATCA[C/T]GAATATAAAAACAACACCCTCTTCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Zfand3
PubMed Links
Gene Details
- Gene
- Zfand3
- Gene Name
- zinc finger, AN1-type domain 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_148926.2 |
|
Intron |
|
|
NP_683728.1 |
View Full Product Details