Product Details

SNP ID
rs13483135
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:83478060 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CGAGGAGCAGAGCGAGTGGGGTAGC[A/G]AAGACCTTGGTGTGGTGATTGATGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Eml4 PubMed Links

Gene Details

Gene
Eml4
Gene Name
echinoderm microtubule associated protein like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114361.1 2808 Missense Mutation NP_001107833.1
NM_001114362.1 2808 Missense Mutation NP_001107834.1
NM_001286567.1 2808 Missense Mutation NP_001273496.1
NM_199466.3 2808 Missense Mutation NP_955760.3

View Full Product Details