Product Details

SNP ID
rs13481757
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:34876589 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AAAGAGAATACCACTGAAGACTGGG[C/T]GGCAACAGCAACCCCCCCCACCCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Prpf4b PubMed Links

Gene Details

Gene
Prpf4b
Gene Name
PRP4 pre-mRNA processing factor 4 homolog B (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013830.2 Intron NP_038858.2

View Full Product Details