Product Details
- SNP ID
-
rs13481617
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:102420293 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTTTAGTCTCAGCAGAGGCGGGA[A/G]GATATCTTGGGTTTGAGGCCTGCCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Lgmn
PubMed Links
Gene Details
- Gene
- Lgmn
- Gene Name
- legumain
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_011175.2 |
|
Intron |
|
|
NP_035305.1 |
View Full Product Details