Product Details

SNP ID
rs13481795
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:45885627 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGCTTATTATCCTTCTTGTACACAT[A/G]AGCTTCTTGTTTGTTTCAGATTTAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Atxn1 PubMed Links

Gene Details

Gene
Atxn1
Gene Name
ataxin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199304.1 Intron NP_001186233.1
NM_001199305.1 Intron NP_001186234.1
NM_009124.6 Intron NP_033150.2

View Full Product Details