Product Details
- SNP ID
-
rs13477168
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:67059461 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATGTCCAAACCAATAAGTTAGGCA[A/C]GTGACTTCTCTTTATTTATTTAAAA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Rsrc1
PubMed Links
Gene Details
- Gene
- Rsrc1
- Gene Name
- arginine/serine-rich coiled-coil 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_025822.3 |
|
Intron |
|
|
NP_080098.1 |
View Full Product Details