Product Details

SNP ID
rs13480955
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:34415729 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGTGCTTCTCATTCTGGATGCAACA[C/T]TGTAACAACTTCTTGCCTAATAGTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Dock2 PubMed Links

Gene Details

Gene
Dock2
Gene Name
dedicator of cyto-kinesis 2
There are no transcripts associated with this gene.

Gene
Fam196b
Gene Name
family with sequence similarity 196, member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025382.2 Intron NP_001020553.1

View Full Product Details