Product Details
- SNP ID
-
rs6306834
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:38345357 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCTCCCTTCCTCTTTTATTTTCAC[A/G]GGAATTTGCTATATTATACGCCATT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
Dgkb
PubMed Links
Gene Details
- Gene
- Dgkb
- Gene Name
- diacylglycerol kinase, beta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178681.4 |
|
Intron |
|
|
NP_848796.2 |
View Full Product Details