Product Details
- SNP ID
-
rs6279639
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:26829487 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTCTGTCTGAATCCTTGCTTTTT[C/T]CCCTTCAATGTTCTTATATGTAATA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Spata16
PubMed Links
Gene Details
- Gene
- Spata16
- Gene Name
- spermatogenesis associated 16
View Full Product Details