Product Details
- SNP ID
-
rs3689181
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:22675394 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCAGAGAGCATGGATCACATGACC[C/A]CTCCTAAGCTCTGCCTTAGCTCCTC
- Phenotype
-
- Polymorphism
- C/A, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Rims1
PubMed Links
Gene Details
- Gene
- Rims1
- Gene Name
- regulating synaptic membrane exocytosis 1
There are no transcripts associated with this gene.
View Full Product Details