Product Details

SNP ID
rs13482122
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.14:31018951 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CACTAGGCCTCTTCATCCTGGCCGC[C/G]GACGCTTCAGGAGCAGAGGAGGTCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
Glt8d1 PubMed Links

Gene Details

Gene
Glt8d1
Gene Name
glycosyltransferase 8 domain containing 1
There are no transcripts associated with this gene.

Gene
Gnl3
Gene Name
guanine nucleotide binding protein-like 3 (nucleolar)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153547.6 181 UTR 5 NP_705775.2
Gene
Pbrm1
Gene Name
polybromo 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081251.1 181 Intron NP_001074720.1
Gene
Snord19
Gene Name
small nucleolar RNA, C/D box 19
There are no transcripts associated with this gene.

Gene
Snord69
Gene Name
small nucleolar RNA, C/D box 69
There are no transcripts associated with this gene.

View Full Product Details