Product Details

SNP ID
rs13481754
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:34214777 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GAAGGGTTTACATGGATGGGTCAGT[A/T]TGACTTTCTTCTGGATTAGGACTAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Slc22a23 PubMed Links

Gene Details

Gene
Slc22a23
Gene Name
solute carrier family 22, member 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033167.3 Intron NP_001028339.1

View Full Product Details