Product Details
- SNP ID
-
rs13477139
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:60038480 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGGTGTGTCTGGAGACAGTGCTGG[G/T]GGAAACTTAGCCGCAGCTGTGACCC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Aadac
PubMed Links
Gene Details
- Gene
- Aadac
- Gene Name
- arylacetamide deacetylase (esterase)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_023383.1 |
589 |
Silent Mutation |
|
|
NP_075872.1 |
View Full Product Details