Product Details

SNP ID
rs13459155
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:87257987 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CGCTGCCATGATGGACATCAGCCCC[A/G]TGGTCATGGGCTCTGGCACCTGGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Mcfd2 PubMed Links

Gene Details

Gene
Mcfd2
Gene Name
multiple coagulation factor deficiency 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139295.3 216 Silent Mutation NP_647456.1
NM_176808.5 216 Silent Mutation NP_789778.1

View Full Product Details