Product Details

SNP ID
rs6226633
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:104645927 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ACCTGCTGGGAGCTTCAATCCACAG[C/T]TTAGCCAGGAATAGGAACTCTGTAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Cwc27 PubMed Links

Gene Details

Gene
Cwc27
Gene Name
CWC27 spliceosome-associated protein homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_026072.1 Intron NP_080348.1

View Full Product Details