Product Details
- SNP ID
-
rs13480682
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:85540810 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAGCTGTGTCTCTTCCAAGGCTCC[A/G]TCCCTCCCGTTCGCTGGGAAAGCCG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Btbd11
PubMed Links
Gene Details
- Gene
- Btbd11
- Gene Name
- BTB (POZ) domain containing 11
View Full Product Details