Product Details

SNP ID
rs13478691
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.6:31410023 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGAAAAGATAGTTTGAGGATTTGAT[A/C]AGGTCAGAATTTGATTCTGATTCCC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Mkln1 PubMed Links

Gene Details

Gene
Mkln1
Gene Name
muskelin 1, intracellular mediator containing kelch motifs
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013791.2 Intron NP_038819.1

View Full Product Details