Product Details
- SNP ID
-
rs13477599
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:21679886 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGACGCAGTGGAACCGCAGGTGTGC[C/T]TTGAGGCTGTTAGGGTAGCGAAAGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Prdm13
PubMed Links
Gene Details
- Gene
- Prdm13
- Gene Name
- PR domain containing 13
View Full Product Details