Product Details

SNP ID
rs13478144
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.5:24321401 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GCAGCTGTAGGACAGTGGCCATGTC[G/T]GCACTTAGCCGGGTTTCCAGCCTGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Kcnh2 PubMed Links

Gene Details

Gene
Kcnh2
Gene Name
potassium voltage-gated channel, subfamily H (eag-related), member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013569.2 3504 Silent Mutation NP_038597.2

View Full Product Details