Product Details

SNP ID
rs3711809
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.5:142885577 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GCCGCTTGCAGTGCTTCAGCATGTC[A/G]GCAAGAGCAGGCATGTACACCACCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
D430018E03Rik PubMed Links

Gene Details

Gene
D430018E03Rik
Gene Name
RIKEN cDNA D430018E03 gene
There are no transcripts associated with this gene.

Gene
Fbxl18
Gene Name
F-box and leucine-rich repeat protein 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033312.3 1842 Silent Mutation NP_001028484.2

View Full Product Details