Product Details

SNP ID
rs13465151
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:130681552 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGCAAAGAAACACACAACTTTTGGT[A/T]TTAGAATATAGCAAATGTAAATTCT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Itpa PubMed Links

Gene Details

Gene
Itpa
Gene Name
inosine triphosphatase (nucleoside triphosphate pyrophosphatase)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025922.2 1097 UTR 3 NP_080198.2
Gene
Slc4a11
Gene Name
solute carrier family 4, sodium bicarbonate transporter-like, member 11
There are no transcripts associated with this gene.

View Full Product Details