Product Details
- SNP ID
-
rs13481574
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:87075701 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTCCTCTTCTTCTTTCTCACCAC[A/G]CCTGCCATCATCATCAACACCATCG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Tmem63c
PubMed Links
Gene Details
- Gene
- Tmem63c
- Gene Name
- transmembrane protein 63c
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_172583.2 |
1577 |
Silent Mutation |
|
|
NP_766171.1 |
View Full Product Details