Product Details

SNP ID
rs13476119
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:134210714 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AACACACTGGTCCTGAAGCTGTCCT[A/G]GGTTTTCCTTCCTGGCTCTGCCCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Adora1 PubMed Links

Gene Details

Gene
Adora1
Gene Name
adenosine A1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008533.3 Intron NP_001008533.1
NM_001039510.2 Intron NP_001034599.1
NM_001282945.1 Intron NP_001269874.1
Gene
Mybph
Gene Name
myosin binding protein H
There are no transcripts associated with this gene.

View Full Product Details