Product Details

SNP ID
rs13476435
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:37429671 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GTATAGCAGGACAACAGCAACTTCC[A/T]GCCTACTTCTGCACTCTGTAGAATG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Rc3h2 PubMed Links

Gene Details

Gene
Rc3h2
Gene Name
ring finger and CCCH-type zinc finger domains 2
There are no transcripts associated with this gene.

Gene
Zbtb26
Gene Name
zinc finger and BTB domain containing 26
There are no transcripts associated with this gene.

Gene
Zbtb6
Gene Name
zinc finger and BTB domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_146253.5 364 Missense Mutation NP_666365.1

View Full Product Details