Product Details

SNP ID
rs13479307
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.7:66698680 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGGTCCTGTCAGGACAGCTCAAGAT[G/T]GCTCCTTTCCACAGTGGCCCCCTGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Asb7 PubMed Links

Gene Details

Gene
Asb7
Gene Name
ankyrin repeat and SOCS box-containing 7
There are no transcripts associated with this gene.

Gene
Lins
Gene Name
lines homolog (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001191001.1 Intron NP_001177930.1
NM_152815.2 Intron NP_690028.2

View Full Product Details